7-Aminoflunitrazepam-d7 (CRM), Cayman Chemical
Supplier: Cayman Chemical Company
Certificates
About this item
An internal standard for the quantification of 7-aminoflunitrazepam by GC- or LC-MS
- This product is qualified as a Certified Reference Material (CRM) that has been manufactured and tested to meet ISO/IEC 17025 and ISO 17034 international standards
- This CRM has been characterized by metrologically valid procedures and may be used as a quantitative analytical reference standard
Specifications
- CAS No.:879894-27-6
- Formula:C₁₆H₇D₇FN₃O
- Storage Temperature:–20 °C
- Description:7-Aminoflunitrazepam-d7
Specifications
Frequently Bought Together