Dextrorphan (tartrate) (CRM), Cayman Chemical
Catalog # 76544-036
Supplier: Cayman Chemical Company
CAS Number:
Specifications
- Concentration:1 mg/ml
- Matrix:Methanol
- Pack type:Glass ampule
- CAS No.:143-98-6
- Description:Dextrorphan (tartrate) (CRM)
- Size:1 mg
- Synonym(s):d-3-hydroxy-N-Methylmorphinan tartrate; NIH 4591
- Formula:C₁₇H₂₃NO·C₄H₆O₆
- Storage Temperature:–20 °C
- Description:Dextrorphan (tartrate)
- Cat. No.:76544-036
Specifications
About this item
A Certified Reference Material
- This product is qualified as a Certified Reference Material (CRM) that has been manufactured and tested to meet ISO/IEC 17025 and ISO 17034 international standards
- This CRM has been characterized by metrologically valid procedures and may be used as a quantitative analytical reference standard