Order Entry
United States
ContactUsLinkComponent
Lorazepam-13C2-d4 (CRM), Cayman Chemical
Lorazepam-13C2-d4 (CRM), Cayman Chemical
Catalog # 76544-024
CAS Number:  
Lorazepam-13C2-d4 (CRM), Cayman Chemical
Catalog # 76544-024
Supplier Number:  18731-100
CAS Number:  

Specifications

  • Concentration:
    100 µg/ml
  • Matrix:
    Acetonitrile
  • Pack type:
    Glass ampule
  • Description:
    Lorazepam-13C2-d4 (CRM)
  • Size:
    100 µg
  • Formula:
    C₁₃[₁₃C]₂H₆D₄Cl₂N₂O₂
  • Storage Temperature:
    –20 °C
  • Description:
    Lorazepam-13C2-d4
  • Cat. No.:
    76544-024

Specifications

About this item

A Certified Reference Material

  • This product is qualified as a Certified Reference Material (CRM) that has been manufactured and tested to meet ISO/IEC 17025 and ISO 17034 international standards
  • This CRM has been characterized by metrologically valid procedures and may be used as a quantitative analytical reference standard