Codeine-d3 (CRM), Cayman Chemical
76543-960
:
- Concentration:1 mg/ml
- Matrix:Methanol
- Pack type:Glass ampule
- CAS No.:70420-71-2
- Description:Codeine-d3 (CRM)
- Size:1 mg
- Synonym(s):Coducept-d3; Methylmorphine-d3; 3-methyl Morphine; Modicept-d3; Morphine-d3 3-methyl ester
- Formula:C₁₈H₁₈D₃NO₃
- Storage Temperature:–20 °C
- Description:Codeine-d3
- Cat. No.:76543-960
A Certified Reference Material
- This product is qualified as a Certified Reference Material (CRM) that has been manufactured and tested to meet ISO/IEC 17025 and ISO 17034 international standards
- This CRM has been characterized by metrologically valid procedures and may be used as a quantitative analytical reference standard