GeneMate Magnetic Stirrer
Product Details & Documents
Magnetic stirrer that is compatible with a wide variety of borosilicate glass beakers, flasks, bottles and other vessels.
- Ceramic work surface, 7.5×7.5"
- Stirring speed from 60 to 1500 rpm
- Space efficient design (8×9") facilitates use on crowded benchtops and/or inside bio-hoods
- Durable, chemical resistant white ceramic work surface
GeneMate magnetic stirrers feature an exceptionally durable, chemical resistant white ceramic work surface. Their space-efficient design (8×9" footprint), makes them ideal for use on crowded bench tops and inside of bio-hoods.
Advanced microprocessor controls with convenient turn knob allow quick, precise adjustment and maintenance of speed. Indicator LED's on the front panel indicate when the stirring function has been activated. With a square 7.5" in work surface, units are compatible with a wide variety of popular sizes of borosilicate glass beakers, flasks, bottles, and other vessels.
Corresponding magnetic hotplate stirrers (stirring and heating 490000-188) or hotplates (heating only 490000-184) with ceramic plates are also available. Full specification details are given under separate entries, please view full details by searching for the relevant product code.
Specifications
- Description:GeneMate Magnetic Stirrer