Masterflex® Polypropylene String-Wound Gradient-Density Cartridges, Avantor®
Supplier: PENTAIR RESIDENTIAL FILTRATION LLC
Certificates
About this item
String-wind pattern has a lower presure drop for higher flow.
- Constructed of polypropylene (PP)
Ideal for removing a broad range of deformable and nondeformable particles such as sand, silt, sludge, rust, and scale. Polypropylene string is resistant to chemical and bacterial attack. Maximum temperature is 150 °F (66 °C).
Note: For optimal performance, change cartridge at 25 psid.
- Environmentally Preferable:
Specifications