Masterflex® Transfer Tubing, Convoluted FEP, Avantor®
Supplier: VWR International
Certificates
About this item
Promotes full drainage.
- Ideal for general-purpose, single-use, and extended fluid path applications
- Global market leader in range of formulations for most extensive chemical compatibility
- Single-use custom lengths and configurations available to minimize downtime
- Regulatory approvals to match your unique application
Convoluted spiral design promotes full drainage and increases tubing flexibility. This chemical-resistant fluoropolymer tubing is sold in 5-ft lengths with nonconvoluted 1"L sections at both ends for easy connections.
- Environmentally Preferable:
Frequently Bought Together