Order Entry
United States
ContactUsLinkComponent
Masterflex® High-Purity Plastic Diaphragm Valves, Avantor®
Masterflex® Valve, PVDF with EPDM Diaphragm, 4.0 Cv
Catalog # MFLX40619-88
Supplier:  VWR International
undefined
Masterflex® Valve, PVDF with EPDM Diaphragm, 4.0 Cv
Catalog # MFLX40619-88
Supplier:  VWR International

Specifications

  • Connection Type:
    Tri-clamp
  • Max. Pressure:
    125 psi
  • Description:
    Valve, PVDF with EPDM Diaphragm, 4.0 Cv
  • Wetted Materials:
    PVDF, EPDM
  • Clamp size:
    0.75"
  • Max. Temperature:
    99 °C (210 °F)
  • Cat. No.:
    MFLX40619-88
  • Cv:
    4
  • Inlet Port:
    ¾" Tri-clamp
  • Outlet Port:
    ¾" tri-clamp

Specifications

About this item

Fully drainable and easy to maintain!

  • Eliminate leaks—ideal for food or pharmaceutical processing
  • Compatible with Tri-Clamp® connections for easy maintenance, change out, and orientation—no cutting or welding needed!
  • Choose either natural polypropylene (PP) or Kynar® (PVDF) body—both are excellent for ultrapure applications
  • Models with Kynar® (PVDF) body offer additional chemical resistance to corrosives

These high-purity valves feature manual actuators. Maximum viscosity is 400 cSt. Maximum fluid temperature is 180 °F (82 °C).

  • Connection Type:
    Tri-clamp
  • Max. Pressure:
    125 psi
  • Description:
    Valve, PVDF with EPDM Diaphragm, 4.0 Cv
  • Environmentally Preferable:
  • Wetted Materials:
    PVDF, EPDM
  • Clamp size:
    0.75"
  • Max. Temperature:
    99 °C (210 °F)
  • Cat. No.:
    MFLX40619-88
  • Cv:
    4
  • Inlet Port:
    ¾" Tri-clamp
  • Outlet Port:
    ¾" tri-clamp