Masterflex® Barbed Check Valves, PVDF and Viton®, Avantor®
Supplier: VWR International
Certificates
About this item
Prevent backflow to protect your sensitive equipment.
- Corrosion- and chemical-resistant PVDF and Viton® construction
- Install in any position
Miniature hosebarb check valves fit securely onto flexible tubing lines to prevent backflow. Autoclavable.
- Environmentally Preferable:
Frequently Bought Together