ContactUsLinkComponent
Boardwalk® Linear Low Density Industrial Can Liners
Boardwalk® Linear Low Density Industrial Can Liners
Catalog # 76540-008
undefined
Boardwalk® Linear Low Density Industrial Can Liners
Catalog # 76540-008
Supplier Number:  BWKG4347G
Specifications
  • Color:
    Gray
  • Description:
    56 gal. industrial can liners
  • Thickness:
    1.3 mil.
  • Cat. No.:
    76540-008
  • W×L:
    43×47"
Specifications
About this item
A quality can liner that provides trusted waste collection at an economical price.
  • Industrial strength with tear and puncture resistance
  • Made from linear low-density polyethylene
  • Sturdy, star-seal bottom helps prevent leaks
  • Individually folded for quick and easy dispensing
Linear, low-density polyethylene provides industrial strength performance. Puncture and tear resistance for added durability. Sturdy, star-seal bottom helps prevent leaks and keep heavier loads inside the bag. Individually folded in carton for quick and easy dispensing.