Masterflex® Double-Open-End Sediment Filter Cartridges, Avantor®
Polypropylene fiber medium is thermally bonded to a rigid core for added durability.
- Constructed of polypropylene (PP)
Ideal for removing a broad range of deformable and nondeformable particles. Rigid design is ideal for filtering high-solids, higher-viscosity fluids that require higher differential pressures. Cartridge imparts no taste, odor, or color. Maximum temperature is 175 °F (79 °C).
Note: For optimal performance, change cartridges at 25 psid.
- Environmentally Preferable:
Frequently Bought Together