ContactUsLinkComponent
HOBO® Data Loggers Self-Describing 6' Stainless Steel Temperature Sensor
HOBO® Data Loggers Self-Describing 6' Stainless Steel Temperature Sensor
Catalog # 470336-314
undefined
HOBO® Data Loggers Self-Describing 6' Stainless Steel Temperature Sensor
Catalog # 470336-314
Supplier Number:  SD-TEMP-SS-06
Specifications
  • Description:
    Air/water/soil temperature sensor, stainless steel, 6' cable length
  • Cat. No.:
    470336-314
  • Accuracy:
    ±0.15 °C from 0...50 °C
  • Range:
    –40...100 °C
Specifications
About this item
Stainless steel temperature sensor with Self-Describing technology for quick, easy set up.
  • Locking jack to ensure sensor is connected and stay connected
  • Food-grade stainless-steel temperature probe
  • Available in 1.8 m (6')
For use with HOBO MX1104 and MX1105 multi-channel data loggers, this food-grade stainless-steel temperature sensor offers integrated strain relief, so the probe is secured to the logger. It features Self-Describing technology to automatically communicate configuration information to the logger, without any programming or extensive user setup.