



Millon's reagent for the detection of tyrosine
Supplier: Thermo Scientific Chemicals
Certificates
About this item
CAS Number:10045-94-0
MDL Number:MFCD00147623
Density:1.4 g/cm³
Storage Temperature:Ambient
UN:2922
ADR:8,II
:GHS05
:GHS06
:GHS08
:GHS09
Specification Test Results
- Appearance:Clear colorless to light yellow Solution
- Density:1.2 to 1.5 kg/l
- Sensitivity:passes test
Specification Test Results
Frequently Bought Together