Order Entry
United States
ContactUsLinkComponent
(S)-(-)-2-Methyl-CBS-oxazaborolidine 1 M in toluene, AcroSeal™
Health
Corrosive
Flammable
Irritant
(S)-(-)-2-Methyl-CBS-oxazaborolidine 1 M in toluene, AcroSeal™
Catalog # TS43603-0500
CAS Number:  
undefined
Health
Corrosive
Flammable
Irritant
(S)-(-)-2-Methyl-CBS-oxazaborolidine 1 M in toluene, AcroSeal™
Catalog # TS43603-0500
Supplier Number:  43603-0500
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Pack type:
    Glass bottle, AcroSeal®
  • Size:
    50 ml
  • Cat. No.:
    TS43603-0500

Specifications

Specification Test Results

  • Appearance:
    Colorless to amber Liquid
  • Strength:
    0.90 to 1.10 M
  • Titration Argentometric:
    ≥85.0 %

Specification Test Results

About this item

 :
 :277.17 g/mol
Boiling Pt:111 °C (760 mmHg)
Density:0.929 g/cm³
Flash Pt:4 °C
Storage Temperature:Ambient
UN:1294
ADR:3,II
 :GHS08
 :GHS05
 :GHS02
 :GHS07
CAS Number:112022-81-8
Formula:C₁₈H₂₀BNO
MDL Number:MFCD00078439