

Rhodamine B 99+%, pure, laser grade
Catalog # TS41900-0010
Supplier: Thermo Scientific Chemicals
CAS Number:
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Pack type:Glass bottle for solids
- Size:1 g
- Cat. No.:TS41900-0010
Specifications
Specification Test Results
- Appearance:Green Crystalline powder or crystals
- Infrared spectrum:Conforms
- HPLC:≥99.0 %
- UV Lambda max:545 nm (in Methanol)
- Fluorescence max:565 nm (in Methanol)
- Optimum lasing:588 nm (in Methanol)
- Molar absorption coefficient:106000 l/(mol.cm) (at Lambda max)
Specification Test Results
About this item
:
:479.02 g/mol
Merck Index:13,08266
Melting Pt:210…211 °C
EINECS:201-383-9
:GHS05
:GHS07
Storage Temperature:Ambient
CAS Number:81-88-9
Formula:C₂₈H₃₁ClN₂O₃
MDL Number:MFCD00011931