Methylthymol blue sodium salt ACS indicator grade
Supplier: Thermo Scientific Chemicals
Certificates
About this item
CAS Number:1945-77-3
:
Formula:C₃₇H₄₀N₂Na₄O₁₃S
MDL Number:MFCD00148905
:844.76 g/mol
Storage Temperature:Ambient
Specification Test Results
- Appearance:Dark green to brown to black Powder
- Infrared spectrum:Conforms
- Visual transition interval:From pH 6.5 (yellow) to pH 8.5 (blue)
- Suitability:(as metal indicator) Passes test
- Clarity of solution:Passes test
Specification Test Results
Frequently Bought Together