Methyl red, pure indicator
Supplier: Thermo Scientific Chemicals
Certificates
About this item
Synonyms: 2-(4-Dimethylaminophenylazo)benzoic acid, 2-((4-(Dimethylamino)phenyl)diazenyl)benzoic acid
CAS Number:493-52-7
:
Formula:C₁₅H₁₅N₃O₂
MDL Number:MFCD00002425
Molecular Weight:269.3 g/mol
Melting Pt:178…182 °C
Merck Index:12,06199
Storage Temperature:Ambient
EINECS:207-776-1
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:Dark red to brown to purple to violet Powder or crystals
- Infrared spectrum:Conforms
- Melting point:178 to 182 °C
- Solubility:(0.1 % in ethanol) Clear
- Specific extinction E (1%/1cm):≥650 (430 nm, in buffer)
- Visual transition interval:from pH 4.2 (pink or red) to pH 6.2 (yellow)
Specification Test Results
Frequently Bought Together