



Crystal violet, pure indicator
Supplier: Thermo Scientific Chemicals
Certificates
About this item
CAS Number:548-62-9
:
Formula:C₂₅H₃₀ClN₃
MDL Number:MFCD00011750
:407.99 g/mol
Merck Index:13,04406
Melting Pt:173 °C (dec.)
Storage Temperature:Ambient
EINECS:208-953-6
UN:3077
ADR:9,III
:GHS07
:GHS08
:GHS05
:GHS09
Specification Test Results
- Appearance:Dark green Crystalline powder
- Infrared spectrum:Conforms
- UV spectrum:Complies
- Sensitivity:(as indicator) Passes test
Specification Test Results
Frequently Bought Together