Order Entry
United States
Orders LinkContactUsLinkComponent
Boron trifluoride diethyl ether complex (∼48% BF₃)

Boron trifluoride diethyl ether complex (∼48% BF₃)

Flammable
Corrosive
Irritant
Health
Boron trifluoride diethyl ether complex (∼48% BF₃)
Certificates
Boron trifluoride diethyl ether complex (∼48% BF₃)
Boron trifluoride diethyl ether complex (∼48% BF₃)

4 Options Available Below

Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Synonyms:  Boron fluoride, ethyl ether complex, Boron trifluoride ethyl etherate, Boron trifluoride diethyl ether, Boron trifluoride etherate, Diethyl ether boron trifluoride, Boron trifluoride diethyl etherate
  • CAS Number:109-63-7
  • Formula:C₄H₁₀BF₃O
  • MDL Number:MFCD00013194
  • Molecular Weight:141.93 g/mol
  • Merck Index:13,01340
  • Boiling Pt:126 °C (760 mmHg)
  • Melting Pt:–60 °C
  • Density:1.12 g/cm³
  • Flash Pt:58 °C
  • Storage Temperature:Ambient
  • EINECS:203-689-8
  • UN:2604
  • ADR:8,I

Documents

Specifications for Product Family

Specification Test Results

  • Appearance:
    Clear colorless to straw yellow Liquid (May darken on storage)
  • Refractive index:
    1.3429 to 1.3449 (20°C, 589 nm)
  • Assay:
    47.5 to 49 % (BF3)
  • GC:
    ≥97.5 %
Product Family Options

Product Information

  • Pack type
    Size
    Availability
    Price
  • Glass bottle for solids
    1 kg
    Log in for availability
    Log in for availability
    Each (1kg)
    Each (1kg) - $134.00
    $134.00
    Each (1kg)
       $134.00

    Supplier Number: 17456-0010
  • Glass bottle
    2.5 kg
    Log in for availability
    Log in for availability
    Each (2,500g)
    Each (2,500g) - $238.00
    $238.00
    Each (2,500g)
       $238.00

    Supplier Number: 17456-0025
  • Glass bottle for solids
    25 g
    Log in for availability
    Log in for availability
    Each (25g)
    Each (25g) - $25.90
    $25.90
    Each (25g)
       $25.90

    Supplier Number: 17456-0250
  • Glass bottle for solids
    100 g
    Log in for availability
    Log in for availability
    Each (100g)
    Each (100g) - $55.90
    $55.90
    Each (100g)
       $55.90

    Supplier Number: 17456-1000

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.