
Bis(2,2,2-trifluoroethyl) (methoxycarbonylmethyl)phosphonate 90%
Catalog # TS37061-0050
Supplier: Thermo Scientific Chemicals
CAS Number:
Specifications
- Pack type:Glass bottle
- Size:5 g
- Cat. No.:TS37061-0050
Specifications
Specification Test Results
- Appearance:Clear orange to dark brown Liquid
- Infrared spectrum:Conforms
- Refractive index:1.3700 to 1.3740 (20°C, 589 nm)
- GC:≥88 %
Specification Test Results
About this item
:
:318.11 g/mol
Density:1.504 g/cm³
Flash Pt:>110 °C
:GHS07
Storage Temperature:Ambient
CAS Number:88738-78-7
Formula:C₇H₉F₆O₅P
MDL Number:MFCD00009901