

Potassium tris(1-pyrazolyl)borohydride 93%
Catalog # TS29601-0050
Supplier: Thermo Scientific Chemicals
CAS Number:
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Pack type:Glass bottle
- Size:5 g
- Cat. No.:TS29601-0050
Specifications
Specification Test Results
- Appearance:White Crystalline powder
- Infrared spectrum:Conforms
- Melting point:182 to 188 °C
- Titration with HClO4:≥91 %
Specification Test Results
About this item
:
:252.13 g/mol
Melting Pt:182…188 °C
Storage Temperature:Ambient
UN:2813
ADR:4.3,III
:GHS07
:GHS02
CAS Number:18583-60-3
:MFCD03414149
Formula:C₉H₁₀BKN₆
MDL Number:MFCD00040346