Fehling's reagent II (L(+)-potassium sodium tartrate in sodium hydroxide solution)
:6381-59-5
:100 °C (760 mmHg)
:1.24 g/cm³
:Ambient
:1824
:8,II
:GHS05
- Appearance:Clear colorless Solution
- Solution contains:346 g C4H4O6KNa.4H2O + 100g NaOH/l H2O
Frequently Bought Together