


Diphenylamine 99+%, Extra Pure indicator grade, redox indicator
Supplier: Thermo Scientific Chemicals
Certificates
About this item
CAS Number:122-39-4
:
Formula:(C₆H₅)₂NH
MDL Number:MFCD00003014
:169.23 g/mol
Merck Index:1303349
Boiling Pt:302 °C (1013 hPa)
Melting Pt:52…54 °C
Flash Pt:152 °C (closed cup)
Storage Temperature:Ambient
EINECS:204-539-4
UN:3077
ADR:9,III
:GHS06
:GHS08
:GHS09
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specification Test Results
- Appearance:White to off-white Crystalline powder or crystals or flakes
- Infrared spectrum:Conforms
- Sulfated ash:≤0.05 %
- GC:≥99.0 %
- Sensitivity:(to nitrate) Passes test
Specification Test Results
Frequently Bought Together