Order Entry
United States
ContactUsLinkComponent
Methylthymol blue sodium salt, water soluble, pure
Methylthymol blue sodium salt, water soluble, pure
Catalog # TS10637-0050
CAS Number:  
undefined
Methylthymol blue sodium salt, water soluble, pure
Catalog # TS10637-0050
Supplier Number:  10637-0050
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Size:
    5 g
  • Cat. No.:
    TS10637-0050

Specifications

Specification Test Results

  • Appearance:
    Dark green to red-brown to black Powder
  • Infrared spectrum:
    Conforms
  • Loss on drying:
    ≤10 % (110°C, 1 h)
  • UV Lambda max:
    435 to 443 nm (in water)
  • Specific absorbance A (1%/1cm):
    ≥120 (at lambda max, in water)

Specification Test Results

About this item

 :
 :844.76 g/mol
Storage Temperature:Ambient
EINECS:217-743-3
CAS Number:1945-77-3
Formula:C₃₇H₄₀N₂Na₄O₁₃S
MDL Number:MFCD00148905