Order Entry
United States
ContactUsLinkComponent
 
beta-Apo-oxytetracycline, can be used as Secondary Standard, Thermo Scientific
  TS25773-1000
 :  
undefined
beta-Apo-oxytetracycline, can be used as Secondary Standard, Thermo Scientific
  TS25773-1000
 :  25773-1000
 :  

 

  • Pack type:
    Glass bottle
  • CAS No.:
    18751-99-0
  • Size:
    100 mg
  • Formula:
    C₂₂H₂₂N₂O₈
  • Description:
    beta-Apo-oxytetracycline
  • Cat. No.:
    TS25773-1000

 

 

CAS No.: 18751-99-0

MDL No.: MFCD00062827
GHS Symbols: GHS08
Melting Point: 200 to 204 °C (D)
Appearance: Yellow-brown to brown Powder
Storage Temperature: Ambient
Signal word: Warning