Schiff reagent solution
Catalog # TS61117-5000
Supplier: Thermo Scientific Chemicals
CAS Number:
Specifications
- Size:500 ml
- Cat. No.:TS61117-5000
Specifications
Specification Test Results
- Appearance:clear colorless solution with a strong odor of sulfur dioxide
Specification Test Results
About this item
:
Density:0.7 g/cm³
Storage Temperature:Ambient
MDL Number:MFCD00081985