

Mitomycin C (2 mg/48 mg NaCl)
Catalog # TS22694-0020
Supplier: Thermo Scientific Chemicals
CAS Number:
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Pack type:Ampoule
- Size:2 mg
- Cat. No.:TS22694-0020
Specifications
Specification Test Results
- Appearance:Blue to violet Powder
- HPLC:≥99 % with regard to the non-formulated material
Specification Test Results
About this item
:
:334.33 g/mol
Melting Pt:>360 °C
Storage Temperature:Refrigerator
UN:2811
ADR:6.1,III
:GHS07
:GHS08
CAS Number:50-07-7
Formula:C₁₅H₁₈N₄O₅
MDL Number:MFCD00078109