Order Entry
United States
Orders LinkContactUsLinkComponent
Phenol 99%, extra pure

Phenol 99%, extra pure

Corrosive
Toxic
Health
Environment
Phenol 99%, extra pure
Certificates
Phenol 99%, extra pure

4 Options Available Below

Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Synonyms:  Phenylalcohol, Carbolic acid, Hydroxybenzene
  • CAS Number:108-95-2
  • Formula:C₆H₅OH
  • MDL Number:MFCD00002143
  • Molecular Weight:94.11 g/mol
  • Merck Index:12,07390
  • Boiling Pt:182 °C (760 mmHg)
  • Melting Pt:39…42 °C
  • Density:1.07 g/cm³
  • Flash Pt:79 °C
  • Storage Temperature:Ambient
  • EINECS:203-632-7
  • UN:1671
  • ADR:6.1,II
  • REACH:01-2119471329-32
Specifications for Product Family

Specification Test Results

  • Appearance:
    Colorless to white Crystals or fused mass
  • Infrared spectrum:
    Conforms
  • Melting point:
    39 to 42 °C
  • Water:
    ≤0.2 %
  • GC:
    ≥98.5 %

Specifications

Product Family Options

Product Information

  • Pack type
    Size
    Availability
    Price
  • Glass bottle for solids
    50 g
    Log in for availability
    Log in for availability
    Each (50g)
    Each (50g) - $38.80
    $38.80
    Each (50g)
       $38.80

    Supplier Number: 14934-0500
    Specifications:
    More
    Specifications
    Cat. No.
    TS14934-0500
  • Glass bottle for solids
    1 kg
    Log in for availability
    Log in for availability
    Each (1kg)
    Each (1kg) - $169.00
    $169.00
    Each (1kg)
       $169.00

    Supplier Number: 14934-0010
    Specifications:
    More
    Specifications
    Cat. No.
    TS14934-0010
  • Glass bottle
    2.5 kg
    Log in for availability
    Log in for availability
    Each (2,500g)
    Each (2,500g) - $250.00
    $250.00
    Each (2,500g)
       $250.00

    Supplier Number: 14934-0025
    Specifications:
    More
    Specifications
    Cat. No.
    TS14934-0025
  • Plastic drum
    5 kg
    Log in for availability
    Log in for availability
    Each (5kg)
    Each (5kg) - $475.00
    $475.00
    Each (5kg)
       $475.00

    Supplier Number: 14934-0051
    Specifications:
    More
    Specifications
    Cat. No.
    TS14934-0051

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.