


Bromine 99.5+% ACS
Supplier: Thermo Scientific Chemicals
Certificates
About this item
CAS Number:7726-95-6
:
Formula:Br₂
MDL Number:MFCD00010896
:159.81 g/mol
Boiling Pt:58.7 °C (760 mmHg)
Melting Pt:–7.2 °C
Density:3.111 g/cm³
UN:1744
ADR:8,I
:GHS06
:GHS05
:GHS09
Storage Temperature:Ambient
Specification Test Results
- Appearance:Dark red-brown Liquid
- Titration Iodometric:≥99.5 %
- Residue after evaporation:≤0.005 %
- Chlorine (Cl):≤0.05 %
- Iodine (I):≤0.001 %
- Nickel (Ni):≤5 ppm
- Heavy metals (as Pb):≤2 ppm
- Sulfur compounds (as S):≤0.001 %
Specification Test Results
Frequently Bought Together