Ammonia solution 28 - 30% in water ACS
Supplier: Thermo Scientific Chemicals
New Product
Certificates
About this item
Synonyms: Ammonia aqueous, Ammonium hydroxide solution, Ammonium hydroxide, Ammonia water
CAS Number:1336-21-6
:7664-41-7
Formula:NH₃
MDL Number:MFCD00066650
Boiling Pt:32…37.7 °C (1013 hPa)
Melting Pt:–57 °C
Density:0.91 g/cm³
Molecular Weight:35.05 g/mol
UN:2672
ADR:8 III
:GHS07
:GHS05
:GHS09
Storage Temperature:Refrigerator
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:Clear colorless Solution (Free from suspended matter or sediment)
- Residue after ignition:≤0.002 %
- Titration with HCl:28.0 to 30.0 % (as NH3)
- Chloride (Cl):≤0.5 ppm
- Nitrate (NO3):≤2 ppm
- Phosphate (PO4):≤2 ppm
- Sulfate (SO4):≤2 ppm
- Iron (Fe):≤0.2 ppm
- Heavy metals (as Pb):≤0.5 ppm
- Carbon dioxide (CO2):≤0.002 %
- KMnO4-reducing substance:Passes test
Specification Test Results
Frequently Bought Together