



Potassium bis(trimethylsilyl)amide 15% (w/w) 0.7 M in toluene, AcroSeal™
Catalog # TS41823-1000
Supplier: Thermo Scientific Chemicals
CAS Number:
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Pack type:Glass bottle, AcroSeal®
- Size:100 ml
- Cat. No.:TS41823-1000
Specifications
Specification Test Results
- Appearance:Clear to slightly hazy colorless to amber Solution
- Titration with HClO4:0.6 to 0.9 M
- Melting point:83 to 86 °C
- GC:≥95 %
Specification Test Results
About this item
:
Flash Pt:7 °C
Storage Temperature:Ambient
EINECS:424-100-2
UN:2920
ADR:8,I
:GHS07
:GHS08
:GHS05
:GHS02
CAS Number:40949-94-8
Formula:C₆H₁₈KNSi₂
MDL Number:MFCD00010330