Order Entry
United States
ContactUsLinkComponent
3,3'-Diaminobenzidine tetrahydrochloride (DAB HCl) 10 mg, tablets for immunohistochemistry
Health
3,3'-Diaminobenzidine tetrahydrochloride (DAB HCl) 10 mg, tablets for immunohistochemistry
Catalog # TS32800-5000
CAS Number:  
undefined
Health
3,3'-Diaminobenzidine tetrahydrochloride (DAB HCl) 10 mg, tablets for immunohistochemistry
Catalog # TS32800-5000
Supplier Number:  32800-5000
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Pack type:
    Plastic bottle
  • Size:
    500 mg
  • Cat. No.:
    TS32800-5000

Specifications

Specification Test Results

  • Appearance:
    Light grayish white Tablet
  • Assay:
    9 to 11 mg/tablet
  • pH:
    6.0 to 7.5

Specification Test Results

About this item

 :
 :360.11 g/mol
 :GHS08
Storage Temperature:Freezer
CAS Number:7411-49-6
Formula:C₁₂H₁₈Cl₄N₄
MDL Number:MFCD00012969