Order Entry
United States
ContactUsLinkComponent
 
 
 
 
N,N-Dimethylformamide 99.8% for peptide synthesis
 
undefined
 
 
 
N,N-Dimethylformamide 99.8% for peptide synthesis

 

  • Appearance:
    Clear liquid
  • Water:
    ≤0.03 % (K.F.)
  • GC:
    ≥99.8 %
  • Color scale:
    ≤10 APHA
  • Free amine:
    ≤0.001 %
  • Acidity (CH3COOH):
    ≤0.001 %

 

Frequently Bought Together