Order Entry
United States
ContactUsLinkComponent
N,N,N',N'-Tetramethylethylenediamine (TEMED) 99.5%, AcroSeal™, purified by redistillation
Irritant
Corrosive
Flammable
N,N,N',N'-Tetramethylethylenediamine (TEMED) 99.5%, AcroSeal™, purified by redistillation
Catalog # TS43383-1000
CAS Number:  
undefined
Irritant
Corrosive
Flammable
N,N,N',N'-Tetramethylethylenediamine (TEMED) 99.5%, AcroSeal™, purified by redistillation
Catalog # TS43383-1000
Supplier Number:  43383-1000
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Pack type:
    Glass bottle, AcroSeal®
  • Size:
    100 ml
  • Cat. No.:
    TS43383-1000

Specifications

Specification Test Results

  • Appearance:
    Clear colorless Liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.4170 to 1.4190 (20°C, 589 nm)
  • GC:
    ≥99.4 %
  • Color scale:
    ≤20 APHA

Specification Test Results

About this item

 :
 :116.21 g/mol
Boiling Pt:120…122 °C (760 mmHg)
Melting Pt:–55 °C
Density:0.77 g/cm³
Flash Pt:17 °C
UN:2372
ADR:3,II
 :GHS07
 :GHS05
 :GHS02
Storage Temperature:Ambient
CAS Number:110-18-9
Formula:C₆H₁₆N₂
MDL Number:MFCD00008335