

Sodium hypochlorite 4-6% available chlorine in aqueous solution
Supplier: Thermo Scientific Chemicals
Certificates
About this item
CAS Number:7681-52-9
:
Formula:ClNaO
MDL Number:MFCD00011120
:74.44 g/mol
Boiling Pt:111 °C (1013 hPa)
Melting Pt:–30…–20 °C
Density:1.083 g/cm³
Storage Temperature:Refrigerator
EINECS:231-668-3
UN:1791
ADR:8,III
:GHS05
:GHS09
Specification Test Results
- Appearance (Color):Clear yellow
- Appearance (Form):Solution
- Titration Iodometric:4.5 to 5.5 %
Specification Test Results
Frequently Bought Together