

tert-Butyl methyl ether 99.8% ACS, meets analytical specifications of Reagent Ph. Eur. for spectroscopy
Catalog # TS42801-0025
Supplier: Thermo Scientific Chemicals
CAS Number:
Specifications
- Pack type:Glass bottle
- Size:2.5 L
- Cat. No.:TS42801-0025
Specifications
Specification Test Results
- Appearance:Clear liquid
- Infrared spectrum:Conforms
- Water:≤0.05 %
- Density:(20/4°C) 0.740 to 0.742
- GC:≥99.75 %
- UV:1 cm cell vs HPLC-grade water
- Color scale:≤10 APHA
- Residue after evaporation:≤0.0003 %
- Peroxides:≤1 ppm (as H2O2)
Specification Test Results
About this item
:
:88.15 g/mol
Boiling Pt:55.2 °C (1013 hPa)
Melting Pt:–110 °C
Density:0.7404 g/cm³
Flash Pt:–28 °C
UN:2398
ADR:3,II
:GHS07
:GHS02
Storage Temperature:Ambient
CAS Number:1634-04-4
Formula:(CH₃)₃COCH₃
MDL Number:MFCD00008812