Dimethyl sulfoxide-[D₆] 99.9 atom % D, AcroSeal™ for NMR spectroscopy
Catalog # TS42694-1000
Supplier: Thermo Scientific Chemicals
CAS Number:
Specifications
- Pack type:Glass bottle, AcroSeal®
- Size:100 ml
- Cat. No.:TS42694-1000
Specifications
Specification Test Results
- Appearance:Clear colorless Liquid
- Water:≤0.03 % (Coulometric)
- D-Enrichment:≥99.85 %
- Sodium sulfate (Na2SO4):≤80 ppm
- Sodium chloride (NaCl):≤200 ppm
- Iron (Fe):≤10 ppm
- Lead (Pb):≤0.5 ppm
Specification Test Results
About this item
:
:84.09 g/mol
Boiling Pt:190 °C (760 mmHg)
Melting Pt:20.2 °C
Density:1.19 g/cm³
Flash Pt:88 °C
Storage Temperature:Ambient
EINECS:218-617-0
CAS Number:2206-27-1
Formula:D₃CS(O)CD₃
MDL Number:MFCD00002090