

EDTA (ethylenediamine tetraacetic acid) 99.5% ACS
Supplier: Thermo Scientific Chemicals
Certificates
About this item
CAS Number:60-00-4
:
Formula:C₁₀H₁₆N₂O₈
MDL Number:MFCD00003541
:292.25 g/mol
Merck Index:13,03546
Boiling Pt:614 °C (1013 hPa)
Melting Pt:220 °C
Flash Pt:>100 °C
Storage Temperature:Ambient
EINECS:200-449-4
REACH:01-2119486399-18
:GHS07
:GHS08
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specification Test Results
- Appearance:White Crystalline powder
- Infrared spectrum:Conforms
- Titration Complexometric:99.4 to 100.6 %
- Residue after ignition:≤0.2 %
- Calcium (Ca):≤10 ppm
- Iron (Fe):≤50 ppm
- Magnesium (Mg):≤5 ppm
- Heavy metals (as Pb):≤10 ppm
- Nitrilotriacetic acid:≤0.1 % ((HOCOCH4)3N)
- Insoluble matter:≤0.005 % (in dilute NH4OH)
Specification Test Results
Frequently Bought Together