L(+)-Ascorbic acid cis- and trans-mixture 98%
Supplier: Thermo Scientific Chemicals
About this item
Synonyms: Antiscorbic vitamin, Ascorbic Acid, L-Ascorbic acid, Vitamin C, (R)-5-((S)-1,2-Dihydroxyethyl)-3,4-dihydroxyfuran-2(5H)-one, Acidum ascorbicum
- CAS Number:50-81-7
- Formula:C₆H₈O₆
- MDL Number:MFCD00064328
- Molecular Weight:176.13 g/mol
- Merck Index:13,00837
- Boiling Pt:55…65 °C (1013 hPa)
- Melting Pt:190…192 °C (dec.)
- Density:0.778 g/cm³
- Flash Pt:–18 °C
- Storage Temperature:Ambient
- EINECS:213-176-0
- UN:3271
- ADR:3,II
- REACH:exempt
3 Options Available Below
Product Details & Documents
Synonyms: Antiscorbic vitamin, Ascorbic Acid, L-Ascorbic acid, Vitamin C, (R)-5-((S)-1,2-Dihydroxyethyl)-3,4-dihydroxyfuran-2(5H)-one, Acidum ascorbicum
- CAS Number:50-81-7
- Formula:C₆H₈O₆
- MDL Number:MFCD00064328
- Molecular Weight:176.13 g/mol
- Merck Index:13,00837
- Boiling Pt:55…65 °C (1013 hPa)
- Melting Pt:190…192 °C (dec.)
- Density:0.778 g/cm³
- Flash Pt:–18 °C
- Storage Temperature:Ambient
- EINECS:213-176-0
- UN:3271
- ADR:3,II
- REACH:exempt
Specifications for Product Family
Specification Test Results
- Appearance:Clear colorless to light yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.3940 to 1.4000 (20°C, 589 nm)
- GC:≥97.5 %
- Solubility:(5 % in water) Passes test
- pH:2.1 to 2.6 (5 % in water)
- Specific optical rotation:+20.5° to +21.5° (20°C, 589 nm) (c=10, H2O)
- Copper (Cu):≤5 ppm
- Iron (Fe):≤2 ppm
- Heavy metals (as Pb):≤10 ppm
- Impurity:≤0.2 % Oxalic acid
Product Family Options
Product Information
- Pack typeSizeAvailabilityPrice
- Supplier Number: 10502-1000
- Supplier Number: 10502-5000
- Supplier Number: 10502-0025
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.



