


Cobalt(II) chloride, anhydrous 97%
Supplier: Thermo Scientific Chemicals
Certificates
About this item
CAS Number:7646-79-9
:
Formula:Cl₂Co
MDL Number:MFCD00010938
:129.84 g/mol
Merck Index:12,02498
Boiling Pt:1049 °C (1013 hPa)
Melting Pt:735 °C
EINECS:231-589-4
UN:3077
ADR:9,III
:GHS07
:GHS08
:GHS09
Storage Temperature:Refrigerator
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specification Test Results
- Appearance:Blue Powder
- Titration Complexometric:≥96.0 %
- Iron (Fe):≤500 ppm
- Lead (Pb):≤100 ppm
- Nickel (Ni):≤2000 ppm
Specification Test Results
Frequently Bought Together