


Sodium perchlorate 99+% for analysis
Supplier: Thermo Scientific Chemicals
Certificates
About this item
CAS Number:7601-89-0
:
Formula:ClNaO₄
MDL Number:MFCD00003480
:122.44 g/mol
Merck Index:13,08726
Melting Pt:482 °C
EINECS:231-511-9
UN:1502
ADR:5.1,II
:GHS03
:GHS07
:GHS08
Storage Temperature:Ambient
Specification Test Results
- Appearance:White Crystalline powder or crystals
- Assay:≥99.0 % (on dry substance)
- Loss on drying:≤2.0 % (130°C, 2 h)
- pH:6.0 to 8.0 (5 % in water)
- Chloride (Cl):≤30 ppm
- Sulfate (SO4):≤20 ppm
- Calcium (Ca):≤200 ppm
- Iron (Fe):≤5 ppm
- Potassium (K):≤500 ppm
- Heavy metals (as Pb):≤5 ppm
- Insoluble matter:≤0.005 % (in water)
Specification Test Results
Frequently Bought Together