Sodium bromide, anhydrous 99+%, Extra Pure
Supplier: Thermo Scientific Chemicals
Certificates
About this item
CAS Number:7647-15-6
:
Formula:NaBr
MDL Number:MFCD00003475
:102.89 g/mol
Merck Index:13,08666
Boiling Pt:1390 °C (760 mmHg)
Melting Pt:755 °C
Storage Temperature:Ambient
EINECS:231-599-9
Specification Test Results
- Appearance:White Crystalline powder
- Titration Argentometric:≥99.0 % (on dry substance)
- Loss on drying:≤1.0 %
- Heavy metals (as Pb):≤10 ppm
Specification Test Results
Frequently Bought Together