



Aniline 99+% ACS
Supplier: Thermo Scientific Chemicals
Certificates
About this item
CAS Number:62-53-3
:
Formula:C₆H₅NH₂
MDL Number:MFCD00007629
:93.13 g/mol
Merck Index:13,00661
Boiling Pt:184 °C (1013 hPa)
Melting Pt:–6.2 °C
Density:1.0213 g/cm³
Flash Pt:76 °C
EINECS:200-539-3
UN:1547
ADR:6.1,II
:GHS06
:GHS08
:GHS05
:GHS09
Storage Temperature:Ambient
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specification Test Results
- Appearance:Clear liquid (May darken to red-brown on storage)
- Infrared spectrum:Conforms
- Refractive index:1.5845 to 1.5875 (20°C, 589 nm)
- GC:≥99.0 %
- Color scale:≤250 APHA
- Residue after ignition:≤0.005 %
- Hydrocarbons:Passes test
- Chlorobenzene (C6H5Cl):≤0.01 %
- Nitrobenzene (C6H5NO2):(Limit about 0.001 %) Passes test
Specification Test Results
Frequently Bought Together