Diethyl ether stabilised, Technical Grade
Metal drum, 5 L
Catalog # TS44426-0050
Supplier: Thermo Scientific Chemicals
Supplier #: 44426-0050
Diethyl ether stabilised, Technical Grade
Catalog # TS44426-0050
Supplier: Thermo Scientific Chemicals
CAS Number: 60-29-7
Jump to:
Product Details & Documents
Synonyms: Ethyl Ether, Ether, 2,2'-Oxydiethane, 2,2'-Oxybisethane, Ethoxyethane, Ethyl ether
- CAS Number:60-29-7
- Formula:(CH₃CH₂)₂O
- MDL Number:MFCD00011646
- Molecular Weight:74.12 g/mol
- Boiling Pt:34.6 °C (760 mmHg)
- Melting Pt:–116 °C
- Density:0.714 g/cm³
- Flash Pt:–45 °C
- Storage Temperature:Ambient
- UN:1155
- ADR:3,I
Specifications
Specification Test Results
- Appearance:Clear colorless Liquid
- Density:0.713 to 0.716 kg/l
- Stabilizer:5 to 10 ppm BHT
Specifications
- Pack type:Metal drum
- Size:5 L
Specifications
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.



