Order Entry
United States
ContactUsLinkComponent
 
Irritant
Corrosive
Trifluoroacetic acid 99+% for spectroscopy
Catalog # TS43416-1000
CAS Number:  
undefined
Irritant
Corrosive
Trifluoroacetic acid 99+% for spectroscopy
Catalog # TS43416-1000
Supplier Number:  43416-1000
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Size:
    100 ml
  • Cat. No.:
    TS43416-1000

Specifications

Specification Test Results

  • Appearance:
    Clear colorless Liquid
  • Water:
    ≤0.05 %
  • UV:
    at 320 nm A: ≤0.03
  • Color scale:
    ≤10 APHA
  • Residue after evaporation:
    ≤0.005 %
  • Titration with NaOH:
    ≥99.0 %

Specification Test Results

About this item

 :
 :114.02 g/mol
Boiling Pt:72 °C (760 mmHg)
Melting Pt:–15 °C
Density:1.535 g/cm³
UN:2699
ADR:8,I
 :GHS07
 :GHS05
Storage Temperature:Ambient
CAS Number:76-05-1
Formula:C₂HF₃O₂
MDL Number:MFCD00004169