


Formic acid 97% ACS
Supplier: Thermo Scientific Chemicals
Certificates
About this item
CAS Number:64-18-6
:
Formula:HCO₂H
MDL Number:MFCD00003297
:46.03
Boiling Pt:101 °C (760 mmHg)
Melting Pt:8 °C
Density:1.22 g/cm³
Flash Pt:50 °C
Storage Temperature:Refrigerator
EINECS:200-579-1
UN:1779
ADR:8,II
:GHS05
:GHS02
:GHS06
Specification Test Results
- Appearance:Clear liquid
- Infrared spectrum:Conforms
- Refractive index:1.3690 to 1.3710 (20°C, 589 nm)
- Color scale:≤15 APHA
- Residue after evaporation:≤0.003 %
- Titration with NaOH:≥96.0 %
- Dilution test:passes test
- Chloride (Cl):≤0.001 %
- Sulfite (SO3):passes test
- Sulfate (SO4):≤0.003 %
- Ammonium (NH4):≤0.005 %
- Iron (Fe):≤0.001 %
- Heavy metals (as Pb):≤0.001 %
- Free acid (CH3COOH):≤0.4 %
Specification Test Results
Frequently Bought Together