

Chloroform-[D₁] 99.8+ atom % D + 0.03% (v/v) TMS for NMR spectroscopy
Supplier: Thermo Scientific Chemicals
Certificates
About this item
CAS Number:865-49-6
:
Formula:CDCl₃
MDL Number:MFCD00000827
:120.38 g/mol
Boiling Pt:60.8 °C (1013 hPa)
Melting Pt:–64 °C
Density:1.5 g/cm³ (20 °C)
Storage Temperature:Refrigerator
EINECS:212-742-4
UN:1888
ADR:6.1,III
:GHS06
:GHS08
Specification Test Results
- Appearance:Clear colorless Liquid
- Water:≤0.01 % (Coulometric)
- D-Enrichment:≥99.80 %
- TMS:0.028 to 0.032 v/v%
Specification Test Results
Frequently Bought Together