Acetone-[D₆] 99.8 atom % D, AcroSeal™ for NMR spectroscopy
Catalog # TS21737-1000
Supplier: Thermo Scientific Chemicals
CAS Number:
Specifications
- Pack type:Glass bottle
- Size:100 ml
- Cat. No.:TS21737-1000
Specifications
Specification Test Results
- Appearance:Clear colorless Liquid
- Water:≤0.03 % (Coulometric)
- D-Enrichment:≥99.75 %
Specification Test Results
About this item
:
:64.03 g/mol
Boiling Pt:55 °C (760 mmHg)
Melting Pt:–93 °C
Density:0.87 g/cm³
Flash Pt:–20 °C
UN:1090
ADR:3,II
:GHS07
:GHS02
Storage Temperature:Ambient
CAS Number:666-52-4
Formula:D₃CC(O)CD₃
MDL Number:MFCD00044635