


Xylene (mixture of isomers) 99+%, Electronic Grade for residue analysis
TS32795-0010
:
- Pack type:Glass bottle
- Size:1 L
- Cat. No.:TS32795-0010
- Appearance:Clear liquid
- Water:≤0.02 %
- GC:≥99.0 % Xylene isomers + Ethylbenzene
- Color scale:≤10 APHA
- Residue after evaporation:≤0.0005 %
- Chloride (Cl):≤2 ppm
- Arsenic (As):≤10 ppb
- Barium (Ba):≤20 ppb
- Copper (Cu):≤10 ppb
- Iron (Fe):≤50 ppb
- Heavy metals (as Pb):≤1 ppm
- Acidity (HCl):≤0.001 %
:
:106.17 g/mol
:13,10136
:140 °C (1013 hPa)
:–34 °C
:0.865 g/cm³
:23 °C
:215-535-7
:1307
:3,III
:GHS02
:GHS07
:GHS08
:Ambient
:1330-20-7
:C₆H₄(CH₃)₂
:MFCD00077264