
2-Formyl-3-thiopheneboronic acid 97%
Supplier: Thermo Scientific Chemicals
Certificates
About this item
CAS Number:4347-31-3
:
Formula:C₅H₅BO₃S
MDL Number:MFCD01075678
Melting Pt:137…193 °C
:155.97 g/mol
:GHS07
Storage Temperature:Refrigerator
Specification Test Results
- Appearance:White to light beige to light brown Powder or crystals
- Infrared spectrum:Conforms
- HPLC:≥96.0 %
- Sulfur (S):≤0.05 %
Specification Test Results
Frequently Bought Together